missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Poxygrid™ Wire Test Tube Racks: 10-13mm
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Brand: Bel-Art™ F18750-0001
Code : NEW
Additional Details : Weight : 0.24760kg
Description
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
Test Tube Rack | |
Blue | |
Epoxy Coated Steel | |
8 x 4.4 x 2.5 in. (20.5 x 11.3 x 6.4cm) | |
8.07 in. | |
4.45 in. | |
2.51 in. |
Autoclavable | |
Tubes (10 to 13 mm) | |
72 | |
20.5 cm | |
11.3 cm | |
6.4 cm | |
13 mm |