missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Poxygrid™ Tube Slant Rack

Product Code. 15530052 Shop All SP Scienceware Products
Change view
Click to view available options
Positions:
8
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions unitSize
15530052 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15530052 Supplier Bel-Art™ Supplier No. F187940028
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Angled rack is useful for keeping tube caps and marked contents easily readable, frequently used reagents at an ergonomic angle or draining excess supernatant fluid from pellets.

Angled rack is useful for keeping tube caps and marked contents easily readable, frequently used reagents at an ergonomic angle or draining excess supernatant fluid from pellets.

  • Holds eight 50mL conical tubes or other vessels up to 30mm in diameter
  • Can be attached to the underside of a shelf using built in mounting points (hardware not included); keeps samples off the bench but immediately available
  • Steel wire is epoxy coated for long life and easy cleaning; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Autoclavable at 121°C (250°F)

Specifications

Autoclavable Not Autoclavable
Color Blue
Holds Tubes (30 mm), Tubes (5 mL)
Material Epoxy-coated Steel
Positions 8
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Conical tubes or other vessels up to 30mm dia.
Height (English) 0.75 in.
Diameter (Metric) Opening 30 mm
Product Type Tube Rack
Shelves 3 Tiers
Height (Metric) 120 cm
Array 2 x 4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.