Learn More
Bel-Art™ Plate Reader
Product Code. 11328763
Change view
Click to view available options
Display Type:
None
Unit Size:
Each
Product Code. Display Type Unit Size
11328763 None Each
1 options
Product Code. 11328763 Supplier Bel-Art™ Supplier No. F378610000
  • ONLINE EXCLUSIVE PRICE
    £454.00
    £226.56 / Pack of 1
    Save £227.44
    50% Off
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tool allows safe examination of the bottom of plates without risking spills on the face and eyes
  • For examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
  • Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
  • Clear deck for resting plates
  • Large, adjustable mirror is perfect for viewing plate underside
  • Mirror magnification is 3X actual plate size
  • Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes
Hemagglutination, Hemagglutination Inhibition, Hemolysis and Aglutination testing
Specifications
Product Type Microtiter Plate Reader
Display Type None
Magnification 3X
Material Polypropylene, Clear Glass Acrylic Deck
For Use With (Application) Hemagglutination, hemagglutination inhibition, hemolysis and agglutination testing
For Use With (Equipment) Microtiter plates