missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Pipet Filler

Product Code. 13137693
Change view
Click to view available options
Pipet Filler, Vikem vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13137693 Pipet Filler, Vikem vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13137693 Supplier Bel-Art™ Supplier No. F378860000

Please to purchase this item. Need a web account? Register with us today!

Squeeze, then release the bulb to draw fluid into the pipet

  • The pipet aid is removed when fluid fills the bulb, and the pipet top quickly covered with a gloved finger to safely and easily control and release fluid
  • The bulb is Vikem™ vinyl with a tapered polyethylene chuck for quick removal when pipet is filled

Specifications

Autoclavable Not Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette
Grip Type Round
Material Vinyl
Model Standard
Number of Channels 1
Sterility Non-sterile
Type Pipette Filler
For Use With (Equipment) Pipet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.