missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ OdoClave™ Deodorant Autoclave Pads

Product Code. 12390469
Change view
Click to view available options
Scent:
Cherry
Clove
Lemon
Pine
Color:
Amber
Brown
Green
Red
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scent Color Unit Size
12390469 Lemon Amber Pack of 100
11976072 Clove Brown Pack of 100
11956072 Cherry Red Pack of 100
11966072 Pine Green Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12390469 Supplier Bel-Art™ Supplier No. H131980003
  • ONLINE EXCLUSIVE PRICE
    £108.00
    £100.93 / Pack of 100
    Save £7.07
    6% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neutralize autoclave odors with a pleasant fragrance

  • Heat-release deodorant works automatically at autoclave temperatures
  • Open pouch and place in bag prior to autoclaving
  • Three-year shelf life or longer if stored in a sealed container
  • 32mm (1.75 in.) square
  • Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads

Order Info

Choose 1.25 in. (32mm) square regular-strength or 2.25 in. (5.7cm) square super-strength pad.

Specifications

Product Type Autoclave Capsule
Color-coded No
Form Liquid
Scent Lemon
Sterility Non-sterile
Autoclavable Autoclavable
Color Amber
Format Pad
For Use With (Application) Autoclave
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.