Learn More
Bel-Art™ SP Scienceware™ No-Wire™ Microcentrifuge Tube Racks
Open sides optimize use in crushed ice or water baths
Brand:  Bel-Art™ F18838-0005
Product Code. 11713197
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Molded polypropylene racks sink in water bath and will not tilt
  • Stackable compact size conserves valuable space
  • Autoclavable at 121°C (250°F)
  • Racks are shipped flat: easy-to-assemble
Order Info
Available in two sizes to accommodate most popular tube sizes. Dimensions (L x W x H): 5 × 4 × 1.7 in. (13 × 11 × 4.3cm)