missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Manomark™ Pens

Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth

Brand:  Bel-Art™ F13377-0000

Code : 03

Additional Details : Weight : 0.08800kg

 View more versions of this product

Product Code. 11715084

  • £69.40 / Pack of 12


In Stock 1
Add to basket

Description

Description

  • Solvent-based ink is hydrocarbon, water and temperature resistant 0°C to 155°C (32°F to 310°F)
  • Marks can be erased with alcohol
  • Unique seal prevents drying out if pen is left uncapped

Specifications

Specifications

Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth
12 Pack
Assorted
Manomark™ Pen
0 to 155°C
Videos