missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Manomark™ Pens

Product Code. 11725084 Shop All SP Scienceware Products
Change view
Click to view available options
Color:
Assorted
Black
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11725084 Black Pack of 12
11715084 Assorted Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11725084 Supplier Bel-Art™ Supplier No. F133770001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth.

  • Solvent-based ink is hydrocarbon, water and temperature resistant 0°C to 155°C (32°F to 310°F)
  • Marks can be erased with alcohol
  • Unique seal prevents drying out if pen is left uncapped

Specifications

Tip Style Felt Tip
Product Type Pen
Color Black
For Use With (Application) Marks on PTFE fluoropolymer resins, glass, wood, plastics, metal, paper, and cloth
Temperature (English) Range 32°F to 310°F
Temperature (Metric) Range 0°C to 155°C
Quantity 12 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.