missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Magnetic Bead Separation Racks
Shop All SP Scienceware Products
Click to view available options
Holds:
PCR Tube Plate (96-well)
Tubes (2 mL)
Tubes (5 or 15 mL)
Unit Size:
Each
Description
- Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays
- Powerful magnets quickly secure beads
- Spring plunger on tube sleeves of 1.5 to 50 mL models allows beads to be sequestered at various heights along the tube wall
- Simply turn tube for efficient bead mixing
- 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes
- Flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells; 6, 12, 24, 48, 96 etc.
- Racks fit microplates following ANSI standards
- Clear acrylic allows unobstructed view of tube contents
Specifications
Specifications
| Product Type | Magnetic Bead Separation Rack |
| Array | 1 x 1 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Holds | PCR Tube Plate (96-well) |
| Material | Neodymium Magnets, Acrylic |
| Positions | 1 |
| Shape | Rectangular |
| Shelves | None |
| Stackable | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction