missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Magnetic Bead Separation Racks

Product Code. 16640584 Shop All SP Scienceware Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Holds:
PCR Tube Plate (96-well)
Tubes (2 mL)
Tubes (5 or 15 mL)
missing translation for 'unitSize'
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
16640584 Tubes (5 or 15 mL) Each
17840839 PCR Tube Plate (96-well) Each
16650584 Tubes (2 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16640584 missing translation for 'mfr' Bel Art™ missing translation for 'supplierNo' F199000001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays.

  • Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves of 1.5 to 50 mL models allows beads to be sequestered at various heights along the tube wall
  • Simply turn tube for efficient bead mixing
  • 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes
  • Flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells; 6, 12, 24, 48, 96 etc.
  • Racks fit microplates following ANSI standards
  • Clear acrylic allows unobstructed view of tube contents

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Tubes (5 or 15 mL)
Material Neodymium Magnets, Acrylic
Positions 6
Shape Cylindrical
Stackable No
Sterility Non-sterile
For Use With (Application) General Laboratories, Immunology, Proteomics, Genomics, Separation Methods
Product Type Magnetic Bead Separation Rack
Shelves None
Height (Metric) 54 mm
Array 1 x 6
Width (Metric) 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.