missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Hand-Held Colony Counter Accessories, Replacement Markers

Product Code. p-4878028 Shop All SP Scienceware Products
Click to view available options
Color:
Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15331252

Brand: Bel-Art™ H378620152

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Bel-Art™ Hand-Held Colony Counter

For use with Hand-Held Colony Counter and System (07-910-018 and 07-910-022).

Specifications

Product Type Replacement Felt Tip Pen
Display Type None
Color Red
For Use With (Application) Works on Plastic or Glass
For Use With (Equipment) Scienceware™ colony counter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.