Learn More
Bel-Art™ SP Scienceware™ Glove Box Clamping Rings
Product Code. 11702156 Shop All SP Scienceware Products
Change view
Click to view available options
For Use With (Equipment):
Fits 6 in. Circular Port Holes
Fits 8 in. Circular Port Holes
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11702156 Fits 8 in. Circular Port Holes Each
11775084 Fits 6 in. Circular Port Holes Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11702156 Supplier Bel-Art™ Supplier No. H500250320
Adjustable rings hold gloves in place
  • Fit Techni-Dome™ glove boxes(Mfr. No. 500300000) or other glove boxes with circular port holes of the correct diameter
Specifications
For Use With (Equipment) Fits 8 in. Circular Port Holes
Product Type Clamping Ring
For Use With (Application) Fasten Glove Firmly to Glove Box