Learn More
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells
Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Brand: Bel-Art™ H40261-0000
Code : Z1
Additional Details : Weight : 1.00000kg
Description
- Made from clear acrylic
- Filler hole screws on each half cavity are fluoropolymer-coated stainless steel
- Volume shown is for one half cell (double for total cell volume)
- A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves
Specifications
Dialysis Cell | |
5 mL | |
Each |
Acrylic | |
Transparent | |
Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket |