missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Desi-Can™ Reusable Desiccant Canister

Product Code. 11812432
Change view
Click to view available options
Diameter (Metric) Outer:
10.1 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
11812432 10.1 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11812432 Supplier Bel-Art™ Supplier No. F420460000
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture, these canisters contain silica gel to effectively absorb moisture through perforations in the aluminum case.

For use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture, these canisters contain silica gel to effectively absorb moisture through perforations in the aluminum case

  • One canister of Reusable Desi-Can Canister maintains a safe level of humidity in 0.09 cubic meter (3 cubic ft.) of space
  • Can be regenerated in 121°C (250°F) oven in about 3 hours for unlimited life
  • Monitor window shows blue when recharged and pink when saturated

Specifications

Diameter (Metric) Outer 10.1 cm
Material Silica Gel
Height (Metric) 1.27 cm
Height (English) 0.5 in.
Product Type Desiccator Canister
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Desi-Can
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Reproductive toxicity Category 1
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H360-May damage fertility or the unborn child.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains cobalt dichloride. May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: cobalt dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.