Learn More
Bel-Art™ SP Scienceware™ Bellows Samplers
Provide quick and easy sampling with one-handed operation
Brand: Bel-Art™ H37823-0050
Code : Z4
Additional Details : CAS Number : 23468-00-0 Weight : 0.17100kg
Description
Provide quick and easy sampling with one-handed operation
- Long tapered tip with 4mm opening
- FDA grade polyethylene construction
- Graduated to indicate volume collected
Specifications
Polyethylene | |
5/Pk. | |
Bag | |
50 cm |
Yes | |
Tapered | |
50 mL | |
Bellows Sampler |