missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Phosphate-free, nonionic
- Will not etch or mar precision glassware
- Rinses quickly; dries without wiping
- Each convenient, pre-portioned pouch makes one gallon of 1% solution
- 3.8 liter (1 gallon) bottle comes with 473ml (16oz) working solution dispenser bottle
- Not for use in automatic dishwashers
Specifications
Specifications
| Product Type | Detergent |
| Color-coded | No |
| Form | Liquid |
| pH | 5 to 9 |
| Scent | Regular Scent |
| Sterility | Non-sterile |
| Boiling Point | >200°C |
| Composition | Ethoxylated Nonylphenol, branched 19 to 21%, Dioxane <0.0006 %, Ethylene Oxide 0.0001%, Water, de |
| Content And Storage | 15°C to 49°C |
| Size | 20 mL |
| Show More |
Safety and Handling
Signal Word
- Warning
- Reproductive toxicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H361-Suspected of damaging fertility or the unborn child.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P202-Do not handle until all safety precautions have been read and understood.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contain: nonylphenol, branched, ethoxylated
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?