missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Aquet™ Detergent

Product Code. 11774133 Shop All SP Scienceware Products
Click to view available options
Volume (English):
0.64 oz.
1 gal.
Unit Size:
Pack of 12
Pack of 20
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11774133

Brand: Bel-Art™ F170940030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For manual cleaning of glassware, plastics and surfaces

  • Phosphate-free, nonionic
  • Will not etch or mar precision glassware
  • Rinses quickly; dries without wiping
  • Each convenient, pre-portioned pouch makes one gallon of 1% solution
  • 3.8 liter (1 gallon) bottle comes with 473ml (16oz) working solution dispenser bottle
  • Not for use in automatic dishwashers

Specifications

Product Type Detergent
Color-coded No
Form Liquid
pH 5 to 9
Scent Regular Scent
Sterility Non-sterile
Boiling Point >200°C
Composition Ethoxylated Nonylphenol, branched 19 to 21%, Dioxane <0.0006 %, Ethylene Oxide 0.0001%, Water, de
Content And Storage 15°C to 49°C
For Use With (Application) For hand washing Glass and Plastic labware
Size 1 gal.
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aquet
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nonylphenol, branched, ethoxylated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.