missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Antibody Saver Trays

Product Code. 12860713
Change view
Click to view available options
Dimensions (L x W):
0.62 x 4.5 in.
6.5 x 6.5 in.
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
12860713 0.62 x 4.5 in. Pack of 5
16695572 6.5 x 6.5 in. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12860713 Supplier Bel Art™ Supplier No. F451000000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Minimize the use of expensive antibodies during incubations by using an optimally sized tray

  • For popular midi and mini gels
  • Stackable, reclosable and reusable
  • All trays include attached lid to protect against spillage
  • Single lane trays are supplied as a group of five joined lanes; can be easily separated to single lane prior to use
  • Polypropylene; autoclavable

Specifications

Capacity (Metric) 1.6 X 11.5 cm per lane
Dimensions (L x W) 0.62 x 4.5 in.
Color Natural
Height (Metric) 25 mm
Length (Metric) 1.6 cm
Material Polypropylene
Type Collection Tray
Width (Metric) 11.5 cm
Includes Attached lid to protect against spillage
Length (English) 0.625 in.
Shape Rectangular
Width (English) 4.5 in.
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.