missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Acetal Plastic Screw Clamp

Complete ON/OFF and fine control with enough accuracy to replace stopcocks in many applications

Brand:  Bel-Art™ 06833-10

Code : A0

Additional Details : Weight : 0.04000kg

Product Code. 11774315

  • £41.80 / Pack of 3
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • Installs without disconnecting tubing or interrupting operation
  • Accepts flexible tubing up to 14mm (0.5 in.) O.D
  • Resistant to acids, alkalis and organic solvents
  • Autoclavable

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers