Learn More
Bel-Art™ SP Scienceware™ 28-Piece Fittings Assortment Kit
Compatible with tubing from 3.2 to 12.7mm (⅛ to ½ in.). Bel Art™ SP Scienceware™ 28-Piece Fittings Assortment Kit are for the most common lab connections.
Brand: Bel-Art™ F19626-0000
Code : 41
Additional Details : Weight : 0.17100kg
Description
- Molded of autoclavable FDA grade polypropylene except quick disconnect fittings which are polyethylene and not autoclavable or FDA grade
- Kit contains: 6 Straight Tubing Connectors, 7 Stepped Tubing Connectors, 5 Wye (Y) Tubing Connectors, 3 “T” shaped Tubing Connectors, 5 Quick Disconnects and 2 flow control Plastic Tubing Clamps
- Plastic organizer box has room for adding other fittings and parts
Specifications
Fittings Kit | |
0.19 to 0.5 in. Tubing |