Learn More
H-B Instrument™ DURAC™ Single Channel Timer with Ten Button Direct Input
Shop All SP Scienceware Products
Click to view available options
:
Traceable to NIST
Description
Includes
Two AAA batteries
- Timing capacity: 99 minutes: 99 seconds
- Count direction: up/down
- Last time set memory
- Audible and visual alarms; displays elapsed time after countdown
- Mounting options: lay flat on bench, flip-open stand for vertical view, pocket clip for attachment to lab coat, magnetic back for metal surface attachment
- Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Specifications
Specifications
| Product Type | Single Channel Timer |
| Accuracy | ±5 seconds/day |
| Resolution | 1 sec. |
| Battery Type | (2) 1.5 V, type AAA or equivalent (included) |
| Includes | Provided with Certificate of Calibration. This certificate indicates calibration using NIST procedures and is in compliance with ANSI/NCSL Z540-1. |
| Quantity | 10/Cs. |