Learn More
H-B Instrument™ DURAC™ Single Channel Three Button Timer with Memory
Shop All SP Scienceware Products
Click to view available options
Quantity:
10/Cs.
Unit Size:
Each
Description
Includes
One AAA battery
- Timing capacity: 99 minutes: 59 seconds
- Count direction: up/down
- Last time set memory
- Audible alarm; displays elapsed time after countdown
- Mounting options: lay flat on bench, stand on edge for vertical view, magnetic back for metal surface attachment
- Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Specifications
Specifications
| Product Type | Single Channel Timer |
| Accuracy | ±5 seconds/day |
| Resolution | 1 sec. |
| Quantity | 10/Cs. |