missing translation for 'onlineSavingsMsg'
Learn More

Bekanamycin, MedChemExpress

Product Code. 16468656
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468656 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468656 Supplier Medchem Express Supplier No. HYB1174
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bekanamycin (Kanamycin B) is an aminoglycoside antibiotic produced by Streptomyces kanamyceticus, against an array of Gram-positive and Gram-negative bacterial strain.

Bekanamycin (Kanamycin B) is an aminoglycoside antibiotic produced by Streptomyces kanamyceticus, against an array of Gram-positive and Gram-negative bacterial strain.

Specifications

Chemical Name or Material Bekanamycin
CAS 4696-76-8
Color White
Purity Grade Notes Research
Molecular Formula C18H37N5O10
Quantity 100 mg
Synonym Kanamycin B
Solubility Information H2O : ≥ 100 mg/mL (206.82 mM) ∣DMSO : 1 mg/mL (2.07 mM; ultrasonic and warming and heat to 80°C)
SMILES O[C@H]1[C@](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@@]3([H])O[C@H](CN)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)([H])O[C@H](CO)[C@@H](O)[C@@H]1N
Molecular Weight (g/mol) 483.51
Formula Weight 483.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.