missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BEGAIN Monoclonal Antibody (OTI4D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527357
This item is not returnable.
View return policy
Description
BEGAIN Monoclonal Antibody for Western Blot
May sustain the structure of the postsynaptic density (PSD). Target protein binds the guanylate kinase domain of PSD-95/SAP90, may play a role in the post-synaptic membrane.
Specifications
| BEGAIN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BUH8 | |
| BEGAIN | |
| Human recombinant protein fragment corresponding to amino acids 374-517 of BEGAIN produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4D9 | |
| Unconjugated | |
| BEGAIN | |
| BEGAIN; BM948371; brain enriched guanylate kinase associated; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated homolog; brain-enriched guanylate kinase-associated protein; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated protein; Gm897; KIAA1446 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57596 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction