missing translation for 'onlineSavingsMsg'
Learn More

Begacestat, MedChemExpress

Product Code. 16441399
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441399 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441399 Supplier Medchem Express Supplier No. HY14175
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Begacestat (GSI-953) is a selective thiophene sulfonamide inhibitor of amyloid precursor protein gamma-secretase (IC50Aβ40=15 nM) for the treatment of Alzheimer's disease.

Begacestat (GSI-953) is a selective thiophene sulfonamide inhibitor of amyloid precursor protein gamma-secretase (IC50Aβ40=15 nM) for the treatment of Alzheimer's disease.

Specifications

Chemical Name or Material Begacestat
CAS 769169-27-9
Color White
Purity Grade Notes Research
Molecular Formula C9H8ClF6NO3S2
Quantity 1 mg
Synonym GSI-953
Solubility Information DMSO : 50 mg/mL (127.64 mM; Need ultrasonic)
SMILES FC(F)(F)C([C@H](NS(=O)(C1=CC=C(Cl)S1)=O)CO)C(F)(F)F
Molecular Weight (g/mol) 391.74
Formula Weight 391.74
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.