missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beflubutamid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Beflubutamid |
| CAS | 113614-08-7 |
| Analyte or Component Names | Beflubutamid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H17 F4 N O2 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C18H17F4NO2/c1-2-16(17(24)23-11-12-6-4-3-5-7-12)25-13-8-9-15(19)14(10-13)18(20,21)22/h3-10,16H,2,11H2,1H3,(H,23,24) |
| SMILES | CCC(Oc1ccc(F)c(c1)C(F)(F)F)C(=O)NCc2ccccc2 |
| IUPAC Name | N-benzyl-2-[4-fluoro-3-(trifluoromethyl)phenoxy]butanamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction