missing translation for 'onlineSavingsMsg'
Learn More

Bedaquiline, MedChemExpress

Product Code. p-200098766
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16468827

Brand: Medchem Express HY14881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bedaquiline (TMC207) is a diarylquinoline drug and inhibits Mycobacterium tuberculosis (Mtb) F1FO-ATP synthase through targeting of both the c- and the ε-subunit. Bedaquiline has uncoupler activity. Bedaquiline is used for the multi-drug resistant tuberculosis.

Bedaquiline (TMC207) is a diarylquinoline drug and inhibits Mycobacterium tuberculosis (Mtb) F1FO-ATP synthase through targeting of both the c- and the ε-subunit. Bedaquiline has uncoupler activity. Bedaquiline is used for the multi-drug resistant tuberculosis.

Specifications

Chemical Name or Material Bedaquiline
CAS 843663-66-1
Color White
Purity Grade Notes Research
Molecular Formula C32H31BrN2O2
Quantity 5 mg
Synonym TMC207 R207910
Solubility Information DMSO : 12.5 mg/mL (22.50 mM; ultrasonic and warming and heat to 60°C)
SMILES BrC1=CC=C(N=C(OC)C([C@H]([C@@](C2=CC=CC3=C2C=CC=C3)(O)CCN(C)C)C4=CC=CC=C4)=C5)C5=C1
Molecular Weight (g/mol) 555.5
Formula Weight 555.5
Percent Purity 97.19%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.