missing translation for 'onlineSavingsMsg'
Learn More

Bedaquiline fumarate, MedChemExpress

Product Code. 16478827
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478827 5 mg 5mg
16446238 10 mg 10mg
16436586 100 mg 100mg
16422367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478827 Supplier Medchem Express Supplier No. HY14881A
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bedaquiline fumarate, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections.

Bedaquiline fumarate, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections.

Specifications

Chemical Name or Material Bedaquiline fumarate
Physical Form Solid
CAS 845533-86-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C36H35BrN2O6
For Use With (Application) COVID-19-immunoregulation
Synonym R403323 TMC207 fumarate R207910 fumarate
Solubility Information DMSO : 100 mg/mL (148.90 mM; Need ultrasonic)
SMILES OC(/C=C/C(O)=O)=O.BrC1=CC=C(N=C(OC)C([C@H]([C@@](C2=CC=CC3=C2C=CC=C3)(O)CCN(C)C)C4=CC=CC=C4)=C5)C5=C1
Molecular Weight (g/mol) 671.58
Formula Weight 671.58
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.