missing translation for 'onlineSavingsMsg'
Learn More

Beclomethasone Dipropionate, MP Biomedicals™

Product Code. 11478941 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11478941

Brand: MP Biomedicals 0215481491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Antiallergic, antiasthmatic, anti-inflammatory
TRUSTED_SUSTAINABILITY

Specifications

CAS 9-8-5534
Color White
Molecular Formula C28H37ClO7
Quantity 1 g
Synonym beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm
InChI Key KUVIULQEHSCUHY-XYWKZLDCSA-N
SMILES CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)Cl)O)C)C)OC(=O)CC
IUPAC Name [2-[(8R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molecular Weight (g/mol) 521.05
PubChem CID 134129500
Formula Weight 521
Physical Form Solid
Chemical Name or Material Beclomethasone Dipropionate
Show More Show Less
compliance-icons
Product Identifier
  • Beclomethasone Dipropionate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Beclomethasone Dipropionate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.