missing translation for 'onlineSavingsMsg'
Learn More

Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals

Product Code. 15597539 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15597539 250 mg 250mg
15587539 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15597539 Supplier Thermo Scientific Acros Supplier No. 460992500
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-8-5534
Molecular Formula C28H37ClO7
Molecular Weight (g/mol) 521.04
InChI Key KUVIULQEHSCUHY-XYWKZLDCSA-N
Synonym beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm
PubChem CID 134129500
IUPAC Name [2-[(8R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMILES CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)Cl)O)C)C)OC(=O)CC

Specifications

Melting Point 208.0°C
Loss on Drying 1% max. (105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 250 mg
Formula Weight 521.04
Percent Purity 97%
Chemical Name or Material Beclomethasone dipropionate
compliance-icons
Product Identifier
  • Beclomethasone dipropionate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.