missing translation for 'onlineSavingsMsg'
Learn More

Beclometasone dipropionate, MedChemExpress

Product Code. 16449855
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449855 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449855 Supplier Medchem Express Supplier No. HY13571A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Beclometasone dipropionate, the prodrug of Beclometasone, is an orally active and potent glucocorticoid recepter agonist. Beclometasone dipropionate acts via a glucocorticoid receptor and suppresses inflammation and hyperproliferation. Beclometasone dipropionate can be used for asthma .

Beclometasone dipropionate, the prodrug of Beclometasone, is an orally active and potent glucocorticoid recepter agonist. Beclometasone dipropionate acts via a glucocorticoid receptor and suppresses inflammation and hyperproliferation. Beclometasone dipropionate can be used for asthma .

Specifications

Chemical Name or Material Beclometasone dipropionate
CAS 9-8-5534
Color White
Purity Grade Notes Research
Molecular Formula C28H37ClO7
Quantity 250 mg
Solubility Information DMSO : ≥ 100 mg/mL (191.92 mM) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES C[C@@]12[C@](C[C@H](C)[C@]2(OC(CC)=O)C(COC(CC)=O)=O)([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C1)=O
Molecular Weight (g/mol) 521.04
Formula Weight 521.04
Percent Purity 99.38%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.