missing translation for 'onlineSavingsMsg'
Learn More

Beclometasone Dipropionate, LoGiCal

Product Code. 30463522
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30463522

Brand: LoGiCal LGCFOR0199.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beclometasone Dipropionate
CAS 9-8-5534
Analyte or Component Names Beclometasone Dipropionate
Shipping Condition Room Temperature
Molecular Formula C28 H37 Cl O7
Certifications/Compliance ISO 9001
Synonym 9-Chloro-11beta-hydroxy-16beta-methyl-3,20-dioxopregna-1,4-diene-17,21-diyl Dipropanoate, Beclometasone Dipropionate, Betamethasone Dipropionate Imp. E (Pharmeuropa), (11β,16β)-9-Chloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate, 9α-Chloro-16β-methylprednisolone 17,21-dipropionate, Aerobec, Aldecin, Aldecin AQ nasal, Anceron, Andion, Beclacin, Beclate, Beclazone, Beclazone 250, Beclazone 50, Beclomet, Beclometasone 17,21-dipropionate, Beclometasone dipropionate, Beclomethasone 17,21-dipropionate, Beclomethasone 17α,21-dipropionate, Beclomethasone dipropionate, Beclotide, Beclotide 100, Becloval, Beclovent, Beclovent Inhaler, Becodisks, Beconase, Beconase AQ, Becotide, Belchlorhinol, Belcoforte, Belcomet, Clenil A, Entyderma, Inalone O, Inalone R, Korbutone, Propaderm, Propaderm Forte, QVAR, Qvar 50, Rino-Clenil, Sanasthmax, Sanasthmyl, Sanasthymyl, Sch 8020W, Vancenase, Vancenase AQ, Vanceril, Vanceril DS, Ventolair, Viarex, Viarox, orBec, AH 15270XX, Betamethasone Dipropionate Impurity E
InChI Formula InChI=1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1
Solution Type Neat
SMILES CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Industry Type Clinical Testing
Molecular Weight (g/mol) 521.04
Quantity 10 mg
Formula Weight 520.2228 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.