missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Beclin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572828
This item is not returnable.
View return policy
Description
Beclin 2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Involved in 2 distinct lysosomal degradation pathways: acts as a regulator of autophagy and as a regulator of G-protein coupled receptors turnover. Regulates degradation in lysosomes of a variety of G-protein coupled receptors via its interaction with GPRASP1/GASP1. [UniProt].
Specifications
| Beclin 2 | |
| Polyclonal | |
| Unconjugated | |
| Becn2 | |
| Bcl-2; BCS2; beclin 1, autophagy related, pseudogene 1; beclin 2; beclin-1 autophagy-related pseudogene 1; Beclin-1-like protein 1; Beclin-2; BECN1L1; BECN1P1; BECN2; Gm104; PFM 2; PFM2; PRDM 5; PRDM5; PRDM5 protein; RGD1563293; similar to Beclin 1 (Coiled-coil myosin-like BCL2-interacting protein) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 441925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| A8MW95 | |
| Becn2 | |
| Beclin 2 antibody for a 16 amino acid peptide near the amino terminus of human Beclin 2. The immunogen is within amino acids 30 - 80 of Beclin 2. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction