missing translation for 'onlineSavingsMsg'
Learn More

Beclin 1, p Ser90 Antibody, Novus Biologicals™

Product Code. 18323493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323493 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323493 Supplier Novus Biologicals Supplier No. NBP313298

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Beclin 1 Polyclonal antibody specifically detects Beclin 1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Beclin 1 (phospho Ser90). The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8678
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.