missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Beclin 1 Antibody Packantibody specifically detects Beclin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
Specifications
| Antigen | Beclin 1 |
| Applications | Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Conjugate | Unconjugated |
| Formulation | See individual datasheets |
| Gene Alias | ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30 |
| Immunogen | See individual datasheets |
| Quantity | 4 Vials |
| Regulatory Status | RUO |
| Research Discipline | Hypoxia, mTOR Pathway |
| Gene ID (Entrez) | 8678 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?