missing translation for 'onlineSavingsMsg'
Learn More

Beclin 1 Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30734635 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734635 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734635 Supplier Novus Biologicals Supplier No. AF5295A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Beclin 1 Polyclonal antibody specifically detects Beclin 1 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin 1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Host Species Sheep
Immunogen E. coli-derived recombinant human Beclin 1, Met12-Gln239, Accession # Q14457
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8678
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.