missing translation for 'onlineSavingsMsg'
Learn More

Beaucage reagent, MedChemExpress

Product Code. 16469845
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469845 250 mg 250mg
16430719 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469845 Supplier Medchem Express Supplier No. HY100951
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Beaucage reagent is found to be potent in causing DNA cleavage.

Beaucage reagent is found to be potent in causing DNA cleavage.

Specifications

Chemical Name or Material Beaucage reagent
CAS 66304-01-6
Color White
Purity Grade Notes Research
Molecular Formula C7H4O3S2
Quantity 250 mg
Solubility Information DMSO : ≥ 100 mg/mL (499.43 mM) ∣H2O : 0.14 mg/mL (0.70 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C12)SS2(=O)=O
Molecular Weight (g/mol) 200.23
Formula Weight 200.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.