missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BEAN Polyclonal Antibody
Description
Immunogen sequence: RHREYEHGYV SDEHTYSRSS RRMRYACSSS EDWPPPLDIS SDGD Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 68%.
The protein encoded by this gene is one of several proteins that interact with NEDD4, a member of a family of ubiquitin-protein ligases. These proteins have PY motifs in common that bind to the WW domains of NEDD4. NEDD4 is developmentally regulated, and is highly expressed in embryonic tissues. Mutations in this gene (i.e., intronic insertions of >100 copies of pentanucleotide repeats including a (TGGAA)n sequence) are associated with spinocerebellar ataxia type 31. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2010].
Specifications
Specifications
| Antigen | BEAN |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BEAN1 |
| Gene Accession No. | Q3B7T3 |
| Gene Alias | BEAN; Bean1; brain expressed, associated with NEDD4, 1; brain-expressed protein associating with Nedd4; Brain-expressed protein associating with Nedd4 homolog; protein BEAN; protein BEAN1; RGD1562003; SCA31 |
| Gene Symbols | BEAN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?