missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ KingFisher™ mL Food Protection Purification System Tubes & Tips

Product Code. 10453273
Click to view available options
Quantity:
240 tests
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10453273

Brand: Applied Biosystems™ 15951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

BeadRetriever™ Tubes & Tips

The tube strips are designed for processing and isolation of target molecules; one sample uses one tube strip which is made up of five tubes. The disposable plastic tips are designed for shielding the magnetic surface from contact with the sample. The BeadRetriever™ System utilize low binding, disposable plastics materials for optimal magnetic particle processing.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
No. of Tests 240 tests
Product Line KingFisher mL
Quantity 240 tests
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.