missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BDY FL VH032

Description
BODIPY FL VH032 is a high-affinity VHL fluorescent probe (Kd = 3.01 nM) with application in time-resolved fluorescence resonance energy-transfer (TR-FRET) assays for high-throughput identification and characterization of VHL ligands. BODIPY FL VH032 consists of a derivative of von Hippel-Lindau (VHL) ligand VH 032 (Cat. No. 6911) and a fluorophore BODIPY FL (Cat. No. 5465) (with excitation peak at 504 nm and emission peak at 520 nm), joined by a polyethylene glycol (PEG) 4 linker. The BODIPY FL VH032 VHL TR-FRET signal is stable through the 90 - 300 min incubation time, and theTR-FRET binding assay is sensitive, selective and resistant to assay interference. BODIPY FL VH032 also exhibits high VHL affinity in a fluorescence polarization (FP) assay. Excitation maximum ∼ 504 nm; emission maximum ∼ 520 nm.
BODIPY® is a registered trademark of Molecular Probes.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-(tert-Butyl)-21-(5,5-difluoro-7,9-dimethyl-5 H-5λ4,6λ4-dipyrrolo[1,2-c:2 ',1 '-f][1,3,2 ]diazaborinin-3-yl)-4,19-dioxo-6,9,12,15-tetraoxa-3,18-diazahenicosanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-ca |
| Quantity | 2 mg |
| Target | Fluorescent Probes |
| Molecular Formula | C46H62BF2N7O9S |
| Purity | 95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?