Learn More
Invitrogen™ BDNF Monoclonal Antibody (1O9E9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA534960
Description
Immunogen sequence: PMKEANIRGQ GGLAYPGVRT HGTLESVNGP KAGSRGLTSL ADTFEHVIEE LLDEDQKVRP NEENNKDADL YTSRVMLSSQ VPLEPPLLFL LEEYKNYLDA ANMSMRVRRH SDPARRGELS VCDSISEWVT AADKKTAVDM SGGTVTVLEK VPVSKGQLKQ YFYETKCNPM GYTKEGCRGI DKRHWNSQCR TTQSYVRALT MDSKKRIGWR FIRIDTSCVC TLTIKRGR.
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
Specifications
| BDNF | |
| Monoclonal | |
| 1.06 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P21237, P23363 | |
| BDNF | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1O9E9 | |
| Unconjugated | |
| BDNF | |
| abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 12064, 24225 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.