missing translation for 'onlineSavingsMsg'
Learn More

BDNF Antibody (584431) [FITC], Novus Biologicals™

Product Code. 30733719 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733719 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733719 Supplier Novus Biologicals Supplier No. FAB31752F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BDNF Monoclonal antibody specifically detects BDNF in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDNF
Applications ELISA
Classification Monoclonal
Clone 584431
Conjugate FITC
Formulation PBS
Gene Alias abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Pro-BDNF, Met1-Arg247, Accession # P23560
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 627
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.