missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDKRB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26672-1-AP-20UL
This item is not returnable.
View return policy
Description
Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.Specifications
| BDKRB1 | |
| Polyclonal | |
| Unconjugated | |
| BDKRB1 | |
| B1 bradykinin receptor, B1BKR, B1R, BDKRB1, BK 1 receptor, BKB1R, BKR1, BRADYB1, bradykinin receptor B1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 623 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P46663 | |
| BDKRB1 | |
| BDKRB1 Fusion Protein Ag24745 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction