missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BDH2 Monoclonal Antibody (OTI2G1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525142
This item is not returnable.
View return policy
Description
BDH2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Dehydrogenase that mediates the formation of 2,5-dihydroxybenzoic acid (2,5-DHBA), a siderophore that shares structural similarities with bacterial enterobactin and associates with LCN2, thereby playing a key role in iron assimilation and homeostasis. Plays a role in susceptibility to bacterial infection by providing an assimilable source of iron that is exploited by pathogenic bacteria. Also acts as a 3-hydroxybutyrate dehydrogenase (PubMed:16380372). [UniProt].
Specifications
| BDH2 | |
| Monoclonal | |
| 1.00 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BUT1 | |
| BDH2 | |
| Full length human recombinant protein of BDH2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2G1 | |
| Unconjugated | |
| BDH2 | |
| 1810026B04Rik; 3-hydroxybutyrate dehydrogenase 2; 3-hydroxybutyrate dehydrogenase type 2; 3-hydroxybutyrate dehydrogenase, type 2; bdh2; dehydrogenase/reductase (SDR family) member 6; dehydrogenase/reductase SDR family member 6; Dhrs6; EFA6R; FLJ13261; hypothetical protein LOC550507; oxidoreductase UCPA; PRO20933; R-beta-hydroxybutyrate dehydrogenase; SDR15C1; Short chain dehydrogenase/reductase family 15C member 1; short chain dehydrogenase/reductase family 15C, member 1; si:dkey-162b23.2; UCPA-OR; UNQ6308; UNQ6308/PRO20933; zgc:110323 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56898 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction