missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £471.00
Specifications
| Antigen | BDH2 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18431532
|
Novus Biologicals
NBP2-32713-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18060984
|
Novus Biologicals
NBP2-32713 |
0.1 mL |
£471.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BDH2 Polyclonal specifically detects BDH2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| BDH2 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 3-hydroxybutyrate dehydrogenase, type 2, dehydrogenase/reductase (SDR family) member 6, Dehydrogenase/reductase SDR family member 6, DHRS6, EC 1.1.1.-, EC 1.1.1.30, EFA6R, FLJ13261, Oxidoreductase UCPA, PRO20933, R-beta-hydroxybutyrate dehydrogenase, SDR15C1,3-hydroxybutyrate dehydrogenase type 2, short chain dehydrogenase/reductase family 15C, member 1, UCPA-OR, UNQ6308 | |
| BDH2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 56898 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EGAKVIATDINESKLQELEKYPGIQTRVLDVTKKKQIDQFANEVERLDVLFNVAGFVHHGTVLDCEEKDWDFS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title