Learn More
BDH1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-15417-100UL
Description
The BDH1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BDH1. This antibody recognizes human, mouse, rat antigen. The BDH1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| BDH1 | |
| 1000 μg/mL | |
| BDH, BDH1, SDR9C1 | |
| 100 μL | |
| 622 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |