missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDCA-2 (CD303) Mouse anti-Human, BV650, Clone: V24-785, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748003
This item is not returnable.
View return policy
Description
BDCA-2 (CD303) Monoclonal antibody specifically detects BDCA-2 (CD303) in Human samples. It is validated for Flow Cytometry
Specifications
| BDCA-2 (CD303) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV650 under optimal conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| V24-785 | |
| Brilliant Violet 650 | |
| CLEC4C; DLEC; HECL; BDCA2; CLECSF7; CLECSF11; PRO34150 | |
| Human BDCA-2 Recombinant Protein | |
| RUO | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction