missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDCA-2 (CD303) Mouse anti-Human, BV421, Clone: V24-785, BD Horizon™
Mouse Monoclonal Antibody
£224.85 - £412.42
Specifications
| Antigen | BDCA-2 (CD303) |
|---|---|
| Clone | V24-785 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15815049
|
BD Biosciences
566428 |
25 Tests |
£228.00 £224.85 / Each Save £3.15 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15805049
|
BD Biosciences
566427 |
100 Tests |
£418.00 £412.42 / Each Save £5.58 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BDCA-2 (CD303) Monoclonal antibody specifically detects BDCA-2 (CD303) in Human samples. It is validated for Flow Cytometry.Specifications
| BDCA-2 (CD303) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Human | |
| Primary |
| V24-785 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2b κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title