missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Red Nucleic Acid Stain has an excitation maximum of 631nm, and is best excited by the red laser
- It has an emission maximum of 651nm
Specifications
Specifications
| For Use With (Application) |
|
| Color | Red |
| Product Type | Red Nucleic Acid Stain |
| Content And Storage | -20°C |
| Quantity | 0.1 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?