missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Rapid Serum Tube
Description
- With Hemogard™ closure
- Reduces contact with blood on stopper and blood splatter during de-capping
- Compatible with major laboratory automation systems
- V-Notch™ Label: Helps align secondary label
- Thrombin based clot activator: Promotes rapid clotting
- DEHP-free gel seperator: Extended analyte stability
- 3 minute spin at 4000 x g (RCF)
Specifications
Specifications
| Product Type | Rapid Serum Tube |
| Additive | Thrombine |
| Material | Plastic |
| Sterility | Sterile |
| Closure Type | Hemogard |
| Capacity (Metric) | 5 mL |
| Closure Color | Orange |
| Tube Size | 13 x 100 mm |
| For Use With (Application) | Serum Determinations, STAT Testing |
| Label Type | Paper |
| Show More |
Safety and Handling
Product Identifier
- TUBE BD VACUTAINER RST 13X100 5.0ML
- Danger
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: protease
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?